Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK gamma Antibody (46B844), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
$206.50 - $499.50
Specifications
Antigen | IKK gamma |
---|---|
Clone | 46B844 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 2 ug/ml, Flow Cytometry 1:10-1:1000 |
Applications | Western Blot, Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056532T
![]() |
Novus Biologicals
NB10056532SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056532
![]() |
Novus Biologicals
NB10056532 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
IKK gamma Monoclonal specifically detects IKK gamma in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
IKK gamma | |
0.5 mg/mL | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Immunology, Innate Immunity | |
NP_001093326 | |
8517 | |
Full-length human IKK gamma protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
46B844 | |
Western Blot 2 ug/ml, Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
FIP-3IP2, FIP3NF-kappa-B essential modifier, Fip3p, I-kappa-B kinase subunit gamma, IkB kinase gamma subunit, IkB kinase subunit gamma, IkB kinase-associated protein 1, IKKAP1, IKKG, IKK-gammaIPD2, incontinentia pigmenti, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma, Inhibitor of nuclear factor kappa-B kinase subunit gamma, IP, IP1, NEMOAMCBX1, NFkappaB essential modulator, NF-kappa-B essential modulator | |
IKBKG | |
IgG1 | |
Protein G purified | |
A 48 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title