Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK gamma Antibody (72C627) - Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233224
Description
IKK gamma Monoclonal specifically detects IKK gamma in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
IKK gamma | |
Monoclonal | |
0.5 mg/mL | |
Western Blot 2-5 μg/mL, Simple Western | |
FIP-3IP2, FIP3NF-kappa-B essential modifier, Fip3p, I-kappa-B kinase subunit gamma, IkB kinase gamma subunit, IkB kinase subunit gamma, IkB kinase-associated protein 1, IKKAP1, IKKG, IKK-gammaIPD2, incontinentia pigmenti, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma, Inhibitor of nuclear factor kappa-B kinase subunit gamma, IP, IP1, NEMOAMCBX1, NFkappaB essential modulator, NF-kappa-B essential modulator | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
A 48 kDa band is observed. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
72C627 | |
Unconjugated | |
Q9Y6K9 | |
IKBKG | |
This antibody was raised against a His-tagged full-length human IKK3/IKKg protein. | |
0.1 mg | |
Apoptosis, Cancer, Immunology, Innate Immunity, Phospho Specific | |
8517 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction