Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Dimer Antibody (28) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200227 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018282 0.025 mg
NBP105045 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018282 Supplier Novus Biologicals Supplier No. NBP1050450.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

D-Dimer Monoclonal specifically detects D-Dimer in Human, Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.

Specifications

Antigen D-Dimer
Applications Western Blot, ELISA
Classification Monoclonal
Clone 28
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:8000
Formulation 10mM Phosphate (pH 7.4) and 0.14M NaCl with 0.05% Sodium Azide
Gene Alias D Dimer
Host Species Mouse
Immunogen Purified human D-dimer adsorbed onto aluminum hydroxide gel.
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Binds human D-dimer. Cross-reacts with directly coated D-monomer but not with directly coated fibrinogen. It also reacts with dog D-dimer.
Target Species Canine, Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.