Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCR9 Antibody (MM0069-6D10) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | CCR9 |
---|---|
Clone | MM0069-6D10 |
Concentration | LYOPH |
Applications | CyTOF, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry |
Classification | Monoclonal |
Description
CCR9 Monoclonal specifically detects CCR9 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
CCR9 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Human | |
C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR-9, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
CCR9 | |
Protein G purified |
MM0069-6D10 | |
CyTOF, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry | |
Unconjugated | |
Mouse | |
Chemokines and Cytokines | |
P51686 | |
10803 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title