Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JAM-B/VE-JAM Antibody (MM0425-4L28) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | JAM-B/VE-JAM |
---|---|
Clone | MM0425-4L28 |
Concentration | LYOPH |
Applications | Blocking Assay, Western Blot |
Classification | Monoclonal |
Description
JAM-B/VE-JAM Monoclonal specifically detects JAM-B/VE-JAM in Human samples. It is validated for Western Blot, Block/Neutralize.Specifications
JAM-B/VE-JAM | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
P57087 | |
58494 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
MM0425-4L28 | |
Blocking Assay, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
C21orf43, CD322 antigen, JAMB, JAM-BJAM-IT/VE-JAM, junctional adhesion molecule 2JAM-2, junctional adhesion molecule B, PRO245, Vascular endothelial junction-associated molecule, VE-JAMVEJAMCD322 | |
JAM2 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title