Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADAM15 Antibody (MM0078-9H34) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
| Antigen | ADAM15 |
|---|---|
| Clone | MM0078-9H34 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
ADAM15 Monoclonal specifically detects ADAM15 in Human samples. It is validated for Western Blot.Specifications
| ADAM15 | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Human | |
| ADAM 15, ADAM metallopeptidase domain 15, disintegrin-like, and cysteine-rich protein 15, EC 3.4.24, MDC-15, Metargidin | |
| ADAM15 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0078-9H34 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q13444 | |
| 8751 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title