Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pref-1/DLK1/FA1 Antibody (MM0514-9D8) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
| Antigen | Pref-1/DLK1/FA1 |
|---|---|
| Clone | MM0514-9D8 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:20-1:200, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF, Western Blot |
Description
Pref-1/DLK1/FA1 Monoclonal specifically detects Pref-1/DLK1/FA1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| Pref-1/DLK1/FA1 | |
| LYOPH | |
| Flow Cytometry, CyTOF, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| delta-like 1 homolog (Drosophila), DLK, DLK-1, FA1fetal antigen 1, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG | |
| DLK1 | |
| Protein G purified |
| MM0514-9D8 | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:20-1:200, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| P80370 | |
| 8788 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title