Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK gamma Antibody - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$182.00 - $464.00
Specifications
Antigen | VEGF |
---|---|
Clone | 6D3 |
Concentration | LYOPH |
Dilution | Western Blot 2-10 ug/ml, ELISA 1-15 ug/ml, Blocking/Neutralizing 50-200 molar excess |
Applications | Western Blot, ELISA, Blocking Assay |
Description
VEGF Monoclonal specifically detects VEGF in Human samples. It is validated for Western Blot, ELISA, Block/Neutralize.Specifications
VEGF | |
LYOPH | |
Western Blot, ELISA, Blocking Assay | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia | |
P15692 | |
7422 | |
IgG1 | |
Protein G purified |
6D3 | |
Western Blot 2-10 ug/ml, ELISA 1-15 ug/ml, Blocking/Neutralizing 50-200 molar excess | |
Monoclonal | |
Purified | |
RUO | |
Human | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
VEGFA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title