Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK gamma Antibody - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$524.00
Specifications
Antigen | AGER |
---|---|
Clone | RM0127-2B65 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:50-1:100 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Description
AGER Monoclonal specifically detects AGER in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
AGER | |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Rat | |
Cancer | |
advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products | |
AGER | |
IgG2 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
RM0127-2B65 | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:50-1:100 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
177 | |
Mouse recombinant protein of the extra cellular domain of RAGE protein | |
Primary | |
This was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse recombinant protein of the extra cellular domain of RAGE protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title