Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK gamma Antibody - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$182.00 - $524.00
Specifications
Antigen | CCL3/MIP-1 alpha |
---|---|
Clone | MM0154-11F16 |
Concentration | LYOPH |
Applications | Immunohistochemistry (Paraffin), Blocking Assay, Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
CCL3/MIP-1 alpha Monoclonal specifically detects CCL3/MIP-1 alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.Specifications
CCL3/MIP-1 alpha | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
P10147 | |
6348 | |
IgG1 | |
Protein G purified |
MM0154-11F16 | |
Immunohistochemistry (Paraffin), Blocking Assay, Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78alpha, Mip1a, MIP-1alpha, MIP1-alpha, Scya3 | |
CCL3 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title