Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ IKKε/TBK1 Inhibitor II, MRT67307

Catalog No. 5063060001
Click to view available options
:
5mg

A cell-permeable, potent, ATP-competitive, and reversible dual kinase inhibitor of TBK1/IKKε (IC50 = 19 and 160 nM, respectively).

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Amber
Description 95% by HPLC
Target Proteins TBK1 kinase dimer interface
Molecular Formula C26H36N6O2
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.