Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKKi/IKK epsilon Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™
SDP

Catalog No. 501993058
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3058 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-3058 Supplier Active Motif Supplier No. 40957
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nuclear factor kappa B (NF-kappa-B) is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kappa-B mediates the expression of a great variety of genes in response to extracellular stimuli. NF-kappa-B is associated with Ikappa-B proteins in the cell cytoplasm, which inhibit NF-kappa-B activity. Ikappa-B is phosphorylated by Ikappa-B kinase (IKK) complex that contains IKK-a, IKK-b, and IKK-gamma. A novel molecule in the IKK complex was recently identified and designated IKK-iota/IKK-i. IKK epsilon is required for the activation of NF-kappa-B by PMA and T cell receptors but not by TNF-a and IL-1. IKK-iota/IKK-i message is expressed in a variety of tissues and is inducible by TNF-a, IL-1, and LPS.

Specifications

Antigen IKKi/IKK epsilon
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.5
Gene IKBKE
Gene Accession No. Q14164
Gene Alias AW558201; I-kappa-B kinase epsilon; IKBKE; Ikke; IKK-E; IKKepsilon; IKK-epsilon; Ikki; IKK-i; IKK-related kinase epsilon; Inducible I kappa-B kinase; inducible IkappaB kinase; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon; inhibitor of kappaB kinase epsilon; Inhibitor of nuclear factor kappa-B kinase subunit epsilon; KIAA0151; RP11-534L20.1
Gene Symbols IKBKE
Host Species Rabbit
Immunogen This Ikki/Ikk epsilon antibody was raised against a synthetic peptide corresponding to amino acid residues 175-188, 525-540, and 567-580 of human Ikki/Ikk epsilon.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9641
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.