Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IKZF3 Monoclonal Antibody (OTI7E11)
GREENER_CHOICE

Catalog No. PIMA526667
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526667 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526667 Supplier Invitrogen™ Supplier No. MA526667
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IKZF3 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This gene product is a transcription factor that is important in the regulation of B lymphocyte proliferation and differentiation. Both Ikaros and Aiolos can participate in chromatin remodeling. Regulation of gene expression in B lymphocytes by Aiolos is complex as it appears to require the sequential formation of Ikaros homodimers, Ikaros/Aiolos heterodimers, and Aiolos homodimers. At least six alternative transcripts encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKZF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ikzf3
Gene Accession No. Q9UKT9
Gene Alias 5830411O07Rik; AIO; AIOLOS; IKAROS family zinc finger 3; IKAROS family zinc finger 3 (Aiolos); ikaros family zinc finger protein 3; Ikzf3; Zfpn1a3; zinc finger DNA binding protein Aiolos; zinc finger protein Aiolos; zinc finger protein, subfamily 1A, 3 (Aiolos); Znfn1a3
Gene Symbols Ikzf3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-301 of IKZF3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22806
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.