Learn More
Invitrogen™ IKZF4 Monoclonal Antibody (GT976)

Description
Positive Control: Raji Predicted Reactivity: Mouse (95%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | IKZF4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | GT976 |
| Concentration | 2.28 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | IKZF4 |
| Gene Accession No. | Q9H2S9 |
| Gene Alias | A630026H08Rik; Eos; IKAROS family zinc finger 4; IKAROS family zinc finger 4 (Eos); ikaros family zinc finger protein 4; Ikzf4; KIAA1782; Zfpn1a4; Zinc finger protein Eos; zinc finger protein, subfamily 1A, 4; zinc finger protein, subfamily 1A, 4 (Eos); zinc finger transcription factor Eos; ZNFN1A4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.