Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IKZF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534963
Description
Recommended positive controls: Raji, K562. Predicted reactivity: Mouse (94%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DNA-binding protein that binds to the 5'GGGAATRCC-3' Ikaros-binding sequence. Transcriptional repressor. Interacts with SPI1 and MITF to repress transcription of the CTSK and ACP5 promoters via recruitment of corepressors SIN3A and CTBP2. May be involved in the development of central and peripheral nervous systems.
Specifications
| IKZF4 | |
| Polyclonal | |
| Unconjugated | |
| IKZF4 | |
| A630026H08Rik; Eos; IKAROS family zinc finger 4; IKAROS family zinc finger 4 (Eos); ikaros family zinc finger protein 4; Ikzf4; KIAA1782; Zfpn1a4; Zinc finger protein Eos; zinc finger protein, subfamily 1A, 4; zinc finger protein, subfamily 1A, 4 (Eos); zinc finger transcription factor Eos; ZNFN1A4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64375 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9H2S9 | |
| IKZF4 | |
| Recombinant protein encompassing a sequence within the center region of human IKZF4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction