Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IKZF4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565433
Description
Immunogen sequence: VNSGGYEKDVE LVAHHSLEPG FGSSLAFVGA EHLRPLRLPP TNCISELTPV ISSVYTQMQP LPGRLELPGS REAGEGPEDL ADGGPLLYRP RGPLTDPGA Highest antigen sequence identity to the following orthologs - mouse 90%, rat 91%.
DNA-binding protein that binds to the 5'GGGAATRCC-3' Ikaros-binding sequence. Transcriptional repressor. Interacts with SPI1 and MITF to repress transcription of the CTSK and ACP5 promoters via recruitment of corepressors SIN3A and CTBP2. May be involved in the development of central and peripheral nervous systems.
Specifications
IKZF4 | |
Polyclonal | |
Unconjugated | |
IKZF4 | |
A630026H08Rik; Eos; IKAROS family zinc finger 4; IKAROS family zinc finger 4 (Eos); ikaros family zinc finger protein 4; Ikzf4; KIAA1782; Zfpn1a4; Zinc finger protein Eos; zinc finger protein, subfamily 1A, 4; zinc finger protein, subfamily 1A, 4 (Eos); zinc finger transcription factor Eos; ZNFN1A4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64375 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H2S9 | |
IKZF4 | |
Recombinant Human IKZF4. Recombinant protein control fragment (Product #RP-104817). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction