Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IKZF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142535
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142535 Supplier Invitrogen™ Supplier No. PA5142535
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognise reported isoforms NP_071910.3; NP_001338019.1; NP_001338020.1; NP_001338021.1. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000.

DNA-binding protein that binds to the 5'GGGAATRCC-3' Ikaros-binding sequence. Transcriptional repressor. Interacts with SPI1 and MITF to repress transcription of the CTSK and ACP5 promoters via recruitment of corepressors SIN3A and CTBP2. May be involved in the development of central and peripheral nervous systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKZF4
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene IKZF4
Gene Accession No. Q8C208, Q9H2S9
Gene Alias A630026H08Rik; Eos; IKAROS family zinc finger 4; IKAROS family zinc finger 4 (Eos); ikaros family zinc finger protein 4; Ikzf4; KIAA1782; Zfpn1a4; Zinc finger protein Eos; zinc finger protein, subfamily 1A, 4; zinc finger protein, subfamily 1A, 4 (Eos); zinc finger transcription factor Eos; ZNFN1A4
Gene Symbols IKZF4
Host Species Goat
Immunogen Peptide with sequence C-RPTFIDRLANSLTKR (aa 309-323).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22781, 64375
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.