Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 alpha/IL-1 beta (Lutikizumab) Antibody - Azide and BSA Free, Novus Biologicals Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP344569
Description
IL-1 alpha/IL-1 beta (Lutikizumab) Monoclonal antibody specifically detects IL-1 alpha/IL-1 beta (Lutikizumab) in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
IL-1 alpha/IL-1 beta (Lutikizumab) | |
Monoclonal | |
Unconjugated | |
Human | |
>90% by SDS-PAGE and HPLC | |
RUO | |
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
Store at 4°C. | |
Purified |
Flow Cytometry, ELISA, Functional Assay | |
LYOPH | |
25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
IL-1 alpha/IL-1 beta (Uniprot: P01583 & P01584) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction