Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IL-1 alpha/IL-1F1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB315SP
This item is not returnable.
View return policy
Description
IL-1 alpha/IL-1F1 Monoclonal specifically detects IL-1 alpha/IL-1F1 in Porcine samples. It is validated for Western Blot.Specifications
IL-1 alpha/IL-1F1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
3552 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
85733 | |
Western Blot 1 ug/mL | |
P18430 | |
IL1A | |
E. coli-derived recombinant porcine IL-1 alpha/IL-1F1 Gln119-Ser270 Accession # P18430 | |
25 μg | |
Primary | |
Detects porcine IL-1 alpha/IL-1F1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 100% cross-reactivity with recombinant human IL-1 alpha , recombinant mouse IL-1 alpha , and recombinant rat IL-1 alpha is observed. | |
Pig | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction