Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-1 alpha Monoclonal Antibody (840C20B8), Biotin
GREENER_CHOICE

Catalog No. AHC0419
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AHC0419 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AHC0419 Supplier Invitrogen™ Supplier No. AHC0419
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is recommended as a detection antibody in Sandwich ELISA applications.

IL-1 alpha (Interleukin-1 alpha) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 alpha is coded by the IL1A gene and signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 beta. IL1A is located on the q arm on chromosome 2 at position 13. IL-1 alpha plays an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses. IL-1 alpha activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 alpha is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. Further, IL-1 alpha is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury and induces apoptosis. The IL1A gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL1 alpha regulates the activities of NF-kappaB and mitogen-activated protein kinases. IL-1 alpha can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of IL-1 alpha are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1 alpha
Applications ELISA
Classification Monoclonal
Clone 840C20B8
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 5mg/mL BSA and 0.5mg/mL sodium azide; pH 7.4
Gene Il1a
Gene Accession No. P01583
Gene Alias CTLA8; hematopoietin-1; IL 1 a; IL 1α; IL1; IL-1 alpha; IL17; IL1A; Il-1a; IL-1alpha; IL1-ALPHA; IL1F1; IL1α; ILN; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin 17; interleukin 1a; interleukin 1-alpha; interleukin 1-alpha (AA 1 - 152); Interleukin1 alpha; interleukin-1 alpha; interleukin-1 alpha precursor; Interleukin-17A; preinterleukin 1 alpha; pre-interleukin-1 alpha; pro-interleukin-1-alpha; RP23-160G19.8
Gene Symbols Il1a
Host Species Mouse
Immunogen Recombinant human IL-1-alpha.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3552
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.