Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 alpha Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991366
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μL
25 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1366 25 μL
50-199-1365 100 μL
50-194-3815 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-199-1366 Supplier Rockland Immunochemicals Supplier No. 209401302S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use.  This is an IgG preparation of whole rabbit serum purified by DEAE fractionation. This antibody is primarily directed against the 17,000 MW human IL-1a and is useful in determining its presence in various assays. In general, this antibody also detects primate IL-1a in the same formats using similar dilutions. The antiserum does not recognize human IL-1b or Mouse or Rabbit IL-1a. In ELISA formats and other immunoreactive assays, this antibody will recognize both the mature 17,000 MW IL-1a as well as the 31,000 MW IL-1a precursor in either non-denatured (native) or denatured samples. Unlike the IL-1b precursor, the native precursor of IL-1a is recognized by the antibody produced to the 17,000 MW form. The 31,000 precursor of IL-1a is biologically active and is found primarily intracellularly. The precursor of IL-1a, unlike that of IL-1b, is biologically active when applied to cells and is thought to have a role as a functional molecule intracellularly and can be found constitutively expressed in various cell. This antibody is also useful for neutralization of human and primate IL-1a activity in bioassays. It does not neutralize the biological activity IL-1b. It does not neutralize the biological activity of mouse, rat or rabbit IL-1a. It will neutralize primate IL-a. For neutralization, it is recommended to incubate the sample with a 1:100 dilution of the antibody for at least 4 hours before being tested. A control of similarly diluted normal rabbit IgG is recommended. This antibody can be used for FACS analysis. Caution should be exhibited as the F( c) domain of the rabbit IgG molecule may interact with cells non-specifically.

IL-1 alpha (Interleukin-1 alpha) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 alpha is coded by the IL1A gene and signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 beta. IL1A is located on the q arm on chromosome 2 at position 13. IL-1 alpha plays an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses. IL-1 alpha activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 alpha is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. Further, IL-1 alpha is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury and induces apoptosis. The IL1A gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL1 alpha regulates the activities of NF-kappaB and mitogen-activated protein kinases. IL-1 alpha can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of IL-1 alpha are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis.

Specifications

Antigen IL-1 alpha
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Il1a
Gene Accession No. P01583
Gene Alias BOS_11618; CTLA8; Hematopoietin-1; IL 1 a; IL 1a; IL1; IL-1 alpha; IL17; IL1A; IL-1A; IL-1alpha; IL1-ALPHA; IL1F1; IL1a; ILN; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin 17; interleukin 1-alpha; Interleukin1 alpha; Interleukin-1 alpha; Interleukin-17A; preinterleukin 1 alpha; pro-interleukin-1-alpha; RP23-160G19.8
Gene Symbols Il1a
Host Species Rabbit
Immunogen The whole rabbit serum used to produce this IgG fraction antibody was prepared by repeated immunizations with recombinant human IL-1a produced in E.coli. The MW of the recombinant IL-1a was 17,000. This is the cleavage site generated by the IL-1b converting enzyme (ICE, capase-1).
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3552
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.