Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 beta/IL-1F2 Antibody (11E5), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073053AF647
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Sandwich ELISA.Specifications
IL-1 beta/IL-1F2 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
IL1B | |
Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant full-length human interleukin 1-beta. | |
0.2 mL | |
Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity, Metastasis | |
3553 | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
1100000 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Sandwich ELISA | |
catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Does not inhibit the biological activity of IL-1. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction