Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 beta/IL-1F2 Antibody (11E5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10073053
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Immunohistochemistry, Sandwich ELISA.Specifications
IL-1 beta/IL-1F2 | |
Monoclonal | |
Unconjugated | |
catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Does not inhibit the biological activity of IL-1. | |
Store at 4C. Do not freeze. | |
IgG1 |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
1100000 | |
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Sandwich ELISA 1:100-1:2000 | |
IL1B | |
Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant full-length human interleukin 1-beta. | |
0.2 mg | |
Adaptive Immunity, Apoptosis, Cancer, Cytokine Research, Diabetes Research, Immunology, Innate Immunity | |
3553 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction