Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 beta/IL-1F2 Antibody (43N1G6), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227342V
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human, Rat samples. It is validated for Western Blot.Specifications
IL-1 beta/IL-1F2 | |
Monoclonal | |
DyLight 405 | |
P01584 | |
IL1B | |
Full-length recombinant human IL-1beta protein was used as immunogen for this antibody. | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity | |
3553 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot | |
43N1G6 | |
Western Blot | |
catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction