Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 beta/IL-1F2 Antibody (8516), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB366701 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB366701 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB366701 Supplier Novus Biologicals Supplier No. FAB201K
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-1 beta/IL-1F2 Monoclonal antibody specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Western Blot, Simple Western, Neutralization, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF

Specifications

Antigen IL-1 beta/IL-1F2
Applications Western Blot, Simple Western, Neutralization, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 8516
Conjugate DyLight 488
Dilution Western Blot, Simple Western, Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-1 beta/IL-1F2, aa 117-269, Accession # P01584
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Cardiovascular Biology, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Lipid and Metabolism, Metastasis
Primary or Secondary Primary
Gene ID (Entrez) 3553
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.