Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IL-1 beta/IL-1F2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB501SP
This item is not returnable.
View return policy
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Rat samples. It is validated for Neutralization, Immunocytochemistry.Specifications
IL-1 beta/IL-1F2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
catabolin, IL-1, IL1 beta, IL-1 beta, IL-1b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3553 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Neutralization, Immunocytochemistry | |
38139 | |
Neutralization 10-30 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q63264 | |
IL1B | |
E. coli-derived recombinant rat IL-1 beta/IL-1F2 Val117-Ser268 Accession # Q63264 | |
25 μg | |
Primary | |
Detects rat IL-1 beta/IL-1F2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human IL-1 alpha , recombinant mouse IL-1 alpha , and recombinant rat IL-1 alpha is observed. | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction