Learn More
Invitrogen™ IL-1 beta Monoclonal Antibody (CRM56), Brilliant Ultra Violet™ 395, eBioscience™, Invitrogen™
Description
Specifications
Specifications
| Antigen | IL-1 beta |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | CRM56 |
| Concentration | 5 μL/Test |
| Conjugate | Brilliant Ultraviolet 395 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | IL1B |
| Gene Accession No. | P01584 |
| Gene Alias | catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.