Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 beta (Pro-form) Monoclonal Antibody (NJTEN3), PE-Cyanine7, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

$211.00 - $474.00

Specifications

Antigen IL-1 beta (Pro-form)
Clone NJTEN3
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-112-3435
Encompass_Preferred
View Documents
Invitrogen™
25711480
25 μg
Each for $211.00
Only null left
Add to Cart
 
50-112-3436
Encompass_Preferred
View Documents
Invitrogen™
25711482
100 μg
Each for $474.00
Only null left
Add to Cart
 
Description

Description

Description: This NJTEN3 monoclonal antibody reacts with the pro-form of mouse IL-1 beta, which is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. It is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is instead secreted alongside caspase-1 via an alternate and incompletely understood mechanism. IL-1 beta signals via the IL-1RI, which is shared with IL-1 alpha. These cytokines play important roles in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses. Their activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions, giving IL-RA or IL-1 beta neutralizing antibodies potential therapeutical value.

IL-1 beta (Pro-form) Monoclonal antibody specifically detects IL-1 beta (Pro-form) in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

IL-1 beta (Pro-form)
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
il 1b; IL 1ß; IL-1 beta; Il1b; Il-1b; IL-1beta; IL1ß; ILN; Interleukin; interleukin 1 beta; Interleukin1 beta; interleukin-1 beta; Interleukin-1b
Il1b
Primary
4°C, store in dark, DO NOT FREEZE!
Il1b
NJTEN3
Flow Cytometry
PE-Cyanine7
Rat
Mouse
P10749
16176
IgG1 κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.