Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 RAcP/IL-1 R3 Antibody (89412), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB676PECY7
Description
IL-1 RAcP/IL-1 R3 Monoclonal antibody specifically detects IL-1 RAcP/IL-1 R3 in Human samples. It is validated for Flow CytometrySpecifications
IL-1 RAcP/IL-1 R3 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
89412 | |
Flow Cytometry | |
C3orf13IL-1RAcPIL1R3Interleukin-1 receptor 3, FLJ37788, IL-1 receptor accessory protein, IL-1R3, IL-1R-3, interleukin 1 receptor accessory protein, interleukin-1 receptor accessory protein, interleukin-1 receptor accessory protein beta | |
S. frugiperda insect ovarian cell line Sf21-derived recombinant human IL-1RAcP/IL-1 R3, Ser21-Glu359 (predicted), Accession # Q9NPH3 | |
0.1 mL | |
Biologically Active Proteins, Cytokine Research, Immunology | |
3556 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction