Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-1 Receptor Type I Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990229
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
100 μL
1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0229 100 μL
50-199-0230 25 μL
50-199-0228 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-199-0229 Supplier Rockland Immunochemicals Supplier No. 1094013040100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. The antiserum is directed against the extracellular domain of the human IL-1R type I, but precipitates the intact receptor expressed on human cells. In general, this antibody also detects primate IL-1RI. The antibody does not recognize human IL-1, IL-1Ra or IL-1R type II. In ELISA formats and other immunoreactive assays, this antibody will detect the extracellular domain of the IL-1R type I (also called soluble IL-1R type I or decoy receptor) found in human body fluids, particularly in the circulation, urine and supernatants of cells and synovial fluid.

The IL-1 system includes two agonists (IL-1 and IL-1), converting enzymes, antagonists, two receptors (IL-1 RI and IL-1 RII) and the IL-1 receptor accessory protein. Interleukin-1 signal is transduced through the type I receptor.

Specifications

Antigen IL-1 Receptor Type I
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 84 mg/mL
Conjugate Unconjugated
Formulation 0.01M sodium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Il1r1
Gene Accession No. P14778
Gene Alias antigen CD121a; CD121 alpha; CD121 antigen-like family member A; CD121a; CD121b; D2S1473; IL-1 receptor alpha chain; IL1R; IL1R1; Il-1r1; IL-1R-1; Il1r-1; Il1ra; IL-1R-alpha; IL1RT1; IL-1RT1; IL-1RT-1; IL-iR; interleukin 1 beta receptor type 1 short transmembrane form; interleukin 1 beta receptor type 1 soluble form; interleukin 1 bete receptor type 1 long transmembrane form; interleukin 1 receptor alpha, type I; interleukin 1 receptor type 1; interleukin 1 receptor, type I; Interleukin-1 receptor alpha; Interleukin-1 receptor type 1; Interleukin-1 receptor type 1, membrane form; Interleukin-1 receptor type 1, soluble form; Interleukin-1 receptor type I; mIL-1R1; p80; sIL-1R1
Gene Symbols Il1r1
Host Species Rabbit
Immunogen IL-1 Receptor Type I Antibody was prepared by repeated immunizations with recombinant human IL-1R type I.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3554
Target Species Human, Non-human Primate
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.