Learn More
IL-1 Receptor Type II Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 109401305
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antiserum is directed against the extracellular domain of the human IL-1R type II, but precipitates the intact receptor expressed on human cells. In general, this antibody also detects primate IL-1RII. The antibody does not recognize human IL-1, IL-1Ra or IL-1R type I. In ELISA formats and other immunoreactive assays, this antibody will detect the extracellular domain of the IL-1R type II (also called soluble IL-1R type II) found in human body fluids, particularly in the circulation, urine and supernatants of cells and synovial fluid.
The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein binds interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA), and acts as a decoy receptor that inhibits the activity of its ligands. Interleukin 4 (IL4) is reported to antagonize the activity of interleukin 1 by inducing the expression and release of this cytokine. This gene and three other genes form a cytokine receptor gene cluster on chromosome 2q12. Two alternatively spliced transcript variants encoding the same protein have been reported.Specifications
| IL-1 Receptor Type II | |
| Polyclonal | |
| Unconjugated | |
| Il1r2 | |
| antigen CDw121b; CD121 antigen-like family member B; CD121b; CDw121b; IL-1 receptor beta chain; IL-1 type II receptor; IL1R2; Il-1r2; IL-1R-2; Il1r-2; IL1R2c; IL1RB; IL-1R-beta; IL-1RT2; IL-1RT-2; ILR2; interleukin 1 receptor type 2; interleukin 1 receptor type II variant 3; interleukin 1 receptor, type II; Interleukin-1 receptor beta; Interleukin-1 receptor type 2; Interleukin-1 receptor type 2, membrane form; Interleukin-1 receptor type 2, soluble form; Interleukin-1 receptor type II; mIL-1R2; sIL-1R2; type II interleukin-1 receptor, beta | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Human, Non-human Primate | |
| Antibody |
| ELISA, Immunoprecipitation, Western Blot | |
| 85 mg/mL | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| P27930 | |
| Il1r2 | |
| This whole rabbit serum was prepared by repeated immunizations with recombinant human IL-1R type II. | |
| RUO | |
| 7850 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.