Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 RI Antibody (007), Alexa Fluor™ 750

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290850AF750
Description
IL-1 RI Monoclonal specifically detects IL-1 RI in Rat samples. It is validated for Flow Cytometry, ELISA.Specifications
IL-1 RI | |
Monoclonal | |
Alexa Fluor 750 | |
CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL-1 RI (NP_037255.3; Met1-Lys352). | |
0.1 mL | |
Cancer, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
3554 | |
Rat | |
Purified |
Flow Cytometry, ELISA | |
7 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Mouse IL1R1 No cross-reactivity in ELISA with: Human IL1R1 Human IL1R3 Human IL1R8 Human IL1R2 Human IL18R1 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction