Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 RI Antibody (129304), Janelia Fluor™ 646, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7712J
This item is not returnable.
View return policy
Description
IL-1 RI Monoclonal antibody specifically detects IL-1 RI in Mouse samples. It is validated for Western Blot, Flow CytometrySpecifications
IL-1 RI | |
Monoclonal | |
Janelia Fluor 646 | |
CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-1 RI, Leu20-Lys338, Accession # P13504 | |
0.5 mL | |
Cancer, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
3554 | |
Store at 4°C in the dark. | |
IgG2b |
Western Blot, Flow Cytometry | |
129304 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction