Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 RI Antibody (JAMA-147), DyLight 488, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NB10065865G
Description
IL-1 RI Monoclonal specifically detects IL-1 RI in Mouse samples. It is validated for Flow Cytometry.Specifications
IL-1 RI | |
Monoclonal | |
DyLight 488 | |
CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
Armenian Hamster | |
Protein G purified | |
RUO | |
Primary | |
NB100-65865 recognizes murine CD121a, which is the 80kD type I interleukin-1 receptor. CD121a is weakly expressed on most cells and tissues including T lymphocytes, thymocytes, epithelial cells and dendritic cells, with stronger expression reported on fibroblasts. The ligands for CD121a are the interleukin-1 (IL-1) family members; IL-1 alpha; IL-1 beta and IL-1 receptor antagonist. IL-1 is a mediator of inflammation and can elicit a variety of other biological responses through binding to CD121a. Clone JAMA-147 has been reported to block IL-1 dependent cellular responses. (1). | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
JAMA-147 | |
Flow Cytometry | |
IL1R1 | |
Extracellular domain of murine IL-1R. | |
0.1 mL | |
Cancer, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
3554 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction