Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 RI Rabbit anti-Rat, Clone: 7, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | IL-1 RI |
---|---|
Clone | 7 |
Dilution | Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000 |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
IL-1 RI Monoclonal specifically detects IL-1 RI in Rat samples. It is validated for Flow Cytometry, ELISA.Specifications
IL-1 RI | |
Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Rat | |
CD121 antigen-like family member A, CD121a antigen, IL1R, IL1R1, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80 | |
Recombinant Rat IL-1 RI protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
7 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Cancer, Cytokine Research, Immunology, Innate Immunity, Phospho Specific | |
0.2 um filtered solution in PBS | |
3554 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title