Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 RII Antibody (20), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289273UV
Description
IL-1 RII Monoclonal antibody specifically detects IL-1 RII in Human samples. It is validated for ELISASpecifications
IL-1 RII | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
20 | |
ELISA | |
beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-1 RII (Met 1-Glu 343; NP_004624.1). | |
0.1 mL | |
Cancer, Phospho Specific, Signal Transduction | |
7850 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction