Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IL-1 RII Antibody Pair [HRP]
SDP

Catalog No. NB009921
Click to view available options
Quantity:
15 Plates

Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-1 RII.

Specifications

Assay Range 9.375 pg/mL
Assay Sensitivity 9.38-600 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-1 RII.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725
Gene ID (Entrez) 7850
Gene Symbol IL1R2
Host Species Mouse/Rabbit
Kit Contents
  • Detection Antibody: 0.5 mg/mL mouse anti-IL-1 RII monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.5 mg/mL of rabbit anti-IL-1 RII monoclonal antibody.
  • Standard: Each vial contains 24 ng of recombinant IL-1 RII.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer, Phospho Specific, Signal Transduction
    Storage Requirements Storage is content dependent.
    Target IL-1 RII
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating 2. Detection Antibody: Dilute to working concentration of 1.0 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 600 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.