Learn More
IL-1 RII Antibody (RM0014-7D7), Alexa Fluor™ 405, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NB11085472AF405
Catalog No. NBT85472405
Only null left
Add to Cart
Description
Description
IL-1 RII Monoclonal specifically detects IL-1 RII in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
IL-1 RII
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
IL1R2
Mouse IL-1 RII (extracellular domain)
0.1 mL
Primary
This recognizes extracellular domain of mouse IL1R-II protein.
Store at 4C in the dark.
IgG2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
RM0014-7D7
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725
Rat
Protein A or G purified
Cancer, Phospho Specific, Signal Transduction
7850
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.