Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1 RII Antibody (RM0014-7D7), Alexa Fluor™ 532, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11085472AF532
Description
IL-1 RII Monoclonal specifically detects IL-1 RII in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IL-1 RII | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
IL1R2 | |
Mouse IL-1 RII (extracellular domain) | |
0.1 mL | |
Primary | |
This recognizes extracellular domain of mouse IL1R-II protein. | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0014-7D7 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725 | |
Rat | |
Protein A or G purified | |
Cancer, Phospho Specific, Signal Transduction | |
7850.0 | |
Mouse | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction