Learn More
                                    IL-10 Antibody (2050B), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB9210MFV450
Description
IL-10 Monoclonal antibody specifically detects IL-10 in Human samples. It is validated for Neutralization, Flow CytometrySpecifications
| IL-10 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
| 3586 | |
| Store at 4C in the dark. | 
| Neutralization, Flow Cytometry | |
| 2050B | |
| Neutralization, Intracellular Staining by Flow Cytometry | |
| CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-10, Met1-Asn178, Accession # P22301 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b | 