Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-10 Chimeric Recombinant Rabbit Monoclonal Antibody (9D7)
GREENER_CHOICE

Catalog No. PIMA548121
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548121 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548121 Supplier Invitrogen™ Supplier No. MA548121
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: The six 9D7 Fab CDR loops interact with a discontinuous IL-10M1 epitope comprised of residues 71-83 and 125-137. 9D7 non-competitively antagonises IL-10R1 binding. Preparation: Rats are immunised with synthetic oligopeptide (containing an N-terminal cysteine)-carrier protein conjugates. IL-10 was conjugated to JES3. The mouse non-secreting myeloma fusion partner P3X63-AG8.653, is fused to immunised rat splenocytes to produce heterohybridomas secreting rat antibody. Bioassays were used to identify neutralising mAbs, and immunochemical screens were used to identify mAbs that bind the cytokine.

IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10 Chimeric
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Neutralization, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 9D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene IL10
Gene Accession No. P22301
Gene Alias CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
Gene Symbols IL10
Host Species Rabbit
Immunogen Rats are immunised with synthetic oligopeptide (containing an N-terminal cysteine)-carrier protein conjugates.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3586
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.