Learn More
Invitrogen™ IL-10 Monoclonal Antibody (67232)

Description
In Western blots, shows approximately 15% cross-reactivity with recombinant mouse IL-10 and less than 2% cross-reactivity with recombinant human IL-10. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | IL-10 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 67232 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | IL10 |
| Gene Accession No. | P29456 |
| Gene Alias | CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.