Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10 Monoclonal Antibody (APM464), Abbomax

Catalog No. 502203829
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3829 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3829 Supplier Abbomax Inc Supplier No. 5008554
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Positive control: IL-10 recombinant protein Cellular location: Secreted.

IL-10 Monoclonal antibody specifically detects IL-10 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)

Specifications

Antigen IL-10
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone APM464
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene IL10
Gene Accession No. P18893, P22301
Gene Alias CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
Host Species Rat
Immunogen A synthetic peptide corresponding to human IL-10 protein
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16153, 3586
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.