Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10 Monoclonal Antibody (JES3-9D7), eBioscience™, Invitrogen™

Rat Monoclonal Antibody

$117.00 - $324.00

Specifications

Antigen IL-10
Clone JES3-9D7
Concentration 0.5 mg/mL
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-136-30
Encompass_Preferred
View Documents
Invitrogen™
14710881
50 μg
Each for $117.00
Only null left
Add to Cart
 
50-136-31
Encompass_Preferred
View Documents
Invitrogen™
14710885
500 μg
Each for $324.00
Only null left
Edge
Add to Cart
 
Description

Description

Description: The JES3-9D7 monoclonal antibody reacts with human interleukin-10 (IL-10). Applications Reported: The JES3-9D7 antibody has been reported for use in capture of human IL-10 by ELISA, intracellular flow cytometric analysis, and for neutralizing IL-10 bioactivity. (Use Functional Grade purified for functional studies cat. 16-7108). Applications Tested: The JES3-9D7 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of human Interleukin-10 (IL-10) in combination with the biotin JES3-12G8 (13-7109) antibody for detection and recombinant human IL-10 (14-8109) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.
Specifications

Specifications

IL-10
0.5 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
IL10
Primary
4°C
IL10
JES3-9D7
ELISA, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rat
Human
P22301
3586
IgG1 κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.