Learn More
Invitrogen™ IL-10 Monoclonal Antibody (JES3-9D7), FITC

Description
IL-10 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | IL-10 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | JES3-9D7 |
| Concentration | 0.1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 4mg/mL BSA and 0.1% sodium azide; pH 7.2 |
| Gene | IL10 |
| Gene Accession No. | P22301 |
| Gene Alias | CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.