Learn More
IL-10 Monoclonal Antibody (JES3-9D7), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$2540.00
Specifications
| Antigen | IL-10 |
|---|---|
| Clone | JES3-9D7 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Functional Assay, Neutralization |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
16-710-838
|
Invitrogen™
16710838 |
5 mg |
Each of 1 for $2,540.00
|
|
|||||
Description
Description: The JES3-9D7 monoclonal antibody reacts with human interleukin-10 (IL-10). Applications Reported: The JES3-9D7 antibody has been reported for use in ELISA capture, ELISPOT capture, neutralization, and intracellular staining for flow cytometric analysis. Applications Tested: The JES3-9D7 antibody has been tested for neutralizing IL-10 binding and biological activity. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.Specifications
| IL-10 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| 3586 | |
| Primary | |
| 4°C | |
| IL10 |
| JES3-9D7 | |
| ELISA, Functional Assay, Neutralization | |
| Functional Grade | |
| Rat | |
| Human | |
| P22301 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.