Learn More
Invitrogen™ IL-10 Monoclonal Antibody (JES5-16E3), Biotin, Functional Grade, eBioscience™

Description
The JES5-16E3 antibody reacts with mouse interleukin-10 (IL-10). Mouse IL-10 is an ~18 kDa factor also known as Cytokine Synthesis Inhibitory Factor (CSIF). In the mouse, Th2 cells, B1 cells, macrophages, and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells, and acts as a costimulatory signal for mast cells, developing thymocytes and the Th2 response.Applications Tested TestedThe Functional Grade Biotin JES5-16E3 antibody has been tested by LAL assay to ensure it is free of endotoxin, and has been tested as the detection antibody in a sandwich ELISA for analysis of mouse IL-10 in combination with the purified JES5-2A5 (14-7102) antibody for capture and recombinant mouse IL-10 (14-8101) as the standard.A suitable range of concentrations of the FG Biotin JES5-16E3 antibody for ELISA detection is 0.5-2 g/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 4000 pg/mL - 30 pg/mL should be included in each ELISA plate.Storage and handling: Use in a sterile environment.
Specifications
Specifications
| Antigen | IL-10 |
| Applications | Functional Assay |
| Classification | Monoclonal |
| Clone | JES5-16E3 |
| Concentration | 0.5 mg/mL |
| Conjugate | Biotin, Functional Grade |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene | IL10 |
| Gene Accession No. | P18893 |
| Gene Alias | CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.