Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-10 Mouse anti-Human, Unconjugated, Clone: 2H6, MilliporeSigma™

Catalog No. MABF22932MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF22932MI 25 μg
MABF22931MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF22932MI Supplier MilliporeSigma Supplier No. MABF229325UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-10 Monoclonal antibody specifically detects IL-10 in Human samples. It is validated for Immunohistochemistry (Paraffin), Affinity Binding Assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2H6
Conjugate Unconjugated
Gene Accession No. NP_000563
Gene Alias Interleukin-10;Cytokine synthesis inhibitory factor;CSIF
Gene Symbols IL10
Host Species Mouse
Immunogen Full-length recombinant mature human IL-10 (without the signal peptide).
Molecular Weight of Antigen 20.52 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 2H6 is a mouse monoclonal antibody that specifically detects human interleukin-10 (IL-10).
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.