Learn More
IL-10 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $1538.97
Specifications
Antigen | IL-10 |
---|---|
Concentration | 85 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0247
![]() ![]() |
Rockland Immunochemicals
109401312S |
25 μL |
Each for $174.73
|
|
|||||
50-199-0246
![]() ![]() |
Rockland Immunochemicals
1094013120100 |
100 μL |
Each for $636.24
|
|
|||||
50-194-3783
![]() ![]() |
Rockland Immunochemicals
109401312 |
1 mL |
Each for $1,538.97
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. The antiserum detects recombinant and native IL-10 present in body fluids and cell supernatants in various assays (ie. IL-1 stimulated IL-10 production from fibroblasts). The antibody recognizes hu IL-10. The antibody will not neutralize the biological activity of murine IL-10.
Interleukin 10 (IL-10, CSIF) is an anti-inflammatory cytokine mainly produced by macrophages and Th2 cells. IL-10 is an 18.6 kDa protein of 160 amino acid residues that shares over 80% sequence homology with the Epstein-Barr Virus protein (BCRFI). The reported biological activities of IL-10, which maybe interrelated, include inhibition of macrophage-mediated cytokine synthesis, suppression of the delayed-type hyper-sensitivity response, and stimulation of the Th2 cell response which results in elevated antibody production. IL-10 functions by inhibiting pro-inflammatory cytokines made by macrophages and regulatory T cells including IFN-gamma, TNF-alpha, IL-2, and IL-3, IL-4, and GM-CSF. IL-10 is also known to suppress antigen presentation on antigen presenting cells, enhances B cell survival, proliferation, and antibody production. IL-10 can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. While human IL-10 can act on murine cells, murine IL-10 cannot act on human cells. Binds to a retinoid X receptor (RXR) responsive element from the cellular retinol-binding protein II promoter (CRBPII-RXRE). IL-10 inhibits the 9-cis-retinoic acid-dependent RXR alpha transcription activation of the retinoic acid responsive element. Knockout studies in mice suggest the function of this cytokine as an essential immunoregulator in the intestinal tract.Specifications
IL-10 | |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P22301 | |
3586 | |
The whole rabbit serum was prepared by repeated immunizations with human IL-10. | |
-20°C, Avoid Freeze/Thaw Cycles | |
IL10 |
85 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
Antibody with 0.01% sodium azide; pH | |
BOS_15514; CSIF; cytokine; Cytokine synthesis inhibitory factor; EGM_01452; GVHDS; H-IL-10; hypothetical protein; IL10; Il-10; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; Interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF | |
IL10 | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.