Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-10 R beta Antibody (MM0360-8R24), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211654PE
Description
IL-10 R beta Monoclonal antibody specifically detects IL-10 R beta in Human samples. It is validated for Western Blot, NeutralizationSpecifications
IL-10 R beta | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Neutralization | |
MM0360-8R24 | |
Western Blot, Block/Neutralize | |
CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL-10R2IL-10R subunit beta, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4 | |
Human recombinant IL-10 R beta (extracellular domain) | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity | |
3588 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction